Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dibutoxy-3-cyclobutene-1,2-dione

Catalog No. 3397925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-979-25G 5 g
33-979-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-979-25G Supplier Sigma Aldrich Supplier No. 3397925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,4-dibutoxy-3-cyclobutene-1,2-dione is the dialkyl ester of squaric acid.3,4-dibutoxy-3-cyclobutene-1,2-dione was used in the synthesis of novel semisquarylium dye, useful for highly selective hg2+ sensing in aqueous media
  • It was also employed in the synthesis of 3-butoxy-4-(1,3,3-trimethyl-2,3-dihydro-1h-2-indolylidenemethyl)-3-cyclobutene-1,2-dione, intermediate for the synthesis of squaryl dyes

Specifications

CAS 2892-62-8
Density 1.047 g/mL (at 25°C (literature))
Boiling Point 139°C (0.5 mmHg)
Molecular Formula C12H18O4
Refractive Index n20/D 1.494 (literature)
Linear Formula [CH3(CH2)3 O]2C4(=O)2
MDL Number MFCD00037150
Quantity 5 g
Synonym Dibutyl squarate
Molecular Weight (g/mol) 226.27
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.