Learn More
Medchemexpress LLC 3,4-dichloro-1-(2,4-dimethoxybenzyl)-1H-pyrrole-2,5-dione | 342416-30-2 | 98.0% | 316.14 g·mol⁻1 | C13H11Cl2NO4 | 5MG
SDP
Catalog No. 5000236434
Encompass_Preferred
IRES-C11 is a small-molecule inhibitor that selectively blocks c-MYC internal ribosome entry site (IRES)-mediated translation by preventing heterogeneous nuclear ribonucleoprotein A1 (hnRNP A1) binding. It reduces c-MYC and cyclin D1 IRES activity in vitro (significant effects observed at ~50 nM) and has shown synergistic anti-glioblastoma effects when combined with mTOR inhibitors. The compound is supplied as a solid powder and as a 10 mM solution in DMSO for research use.
  • Selective inhibitor of c-MYC IRES-mediated translation.
  • Blocks hnRNP A1 interaction with the c-MYC IRES.
  • Reduces c-MYC and cyclin D1 protein levels at nanomolar concentrations.
  • Demonstrates synergy with mTOR inhibitors in glioblastoma models.
  • Available as powder and as a 10 mM solution in DMSO for in vitro studies.
  • High purity (98.01%) and molecular weight 316.14 g·mol⁻1.
Catalog No. 50-002-36434 Supplier Medchemexpress LLC Supplier No. HY1248115MG
IRES-C11 is a small-molecule inhibitor that selectively blocks c-MYC internal ribosome entry site (IRES)-mediated translation by preventing heterogeneous nuclear ribonucleoprotein A1 (hnRNP A1) binding. It reduces c-MYC and cyclin D1 IRES activity in vitro (significant effects observed at ~50 nM) and has shown synergistic anti-glioblastoma effects when combined with mTOR inhibitors. The compound is supplied as a solid powder and as a 10 mM solution in DMSO for research use.
  • Selective inhibitor of c-MYC IRES-mediated translation.
  • Blocks hnRNP A1 interaction with the c-MYC IRES.
  • Reduces c-MYC and cyclin D1 protein levels at nanomolar concentrations.
  • Demonstrates synergy with mTOR inhibitors in glioblastoma models.
  • Available as powder and as a 10 mM solution in DMSO for in vitro studies.
  • High purity (98.01%) and molecular weight 316.14 g·mol⁻1.