Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dichloro-1-(2,4-dimethoxybenzyl)-1H-pyrrole-2,5-dione | 342416-30-2 | 98.0% | 316.14 g/mol | C13H11Cl2NO4 | 10MM 1ML
SDP

Catalog No. 5000239233
Encompass_Preferred

IRES-C11 is a small-molecule inhibitor that selectively targets the internal ribosome entry site (IRES) of c-MYC mRNA, blocking IRES-dependent translation and affecting pathways related to apoptosis and cell proliferation. It is provided in a prepared solution format for in vitro research use.

  • Specific inhibitor of c-MYC IRES-mediated translation.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • Purity 98.0% (HPLC).
  • Molecular weight 316.14 g/mol; formula C13H11Cl2NO4.
  • Intended for in vitro studies of translation regulation and apoptosis.
  • Store according to solvent and powder recommendations for stability.

Catalog No. 50-002-39233 Supplier Medchemexpress LLC Supplier No. HY12481110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

IRES-C11 is a small-molecule inhibitor that selectively targets the internal ribosome entry site (IRES) of c-MYC mRNA, blocking IRES-dependent translation and affecting pathways related to apoptosis and cell proliferation. It is provided in a prepared solution format for in vitro research use.

  • Specific inhibitor of c-MYC IRES-mediated translation.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • Purity 98.0% (HPLC).
  • Molecular weight 316.14 g/mol; formula C13H11Cl2NO4.
  • Intended for in vitro studies of translation regulation and apoptosis.
  • Store according to solvent and powder recommendations for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.