Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichloro-L-phenylalanine, 98%, Thermo Scientific Chemicals

Catalog No. AAH6383603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6383603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6383603 Supplier Thermo Scientific Chemicals Supplier No. H6383603

CAS: 52794-99-7 | Molecular Formula: C9H9Cl2NO2 | Molecular Weight: 234.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52794-99-7
Molecular Formula C9H9Cl2NO2
Molecular Weight (g/mol) 234.076
MDL Number MFCD00191203
InChI Key NRCSJHVDTAAISV-QMMMGPOBSA-N
Synonym s-2-amino-3-3,4-dichlorophenyl propanoic acid, l-3,4-dichlorophenylalanine, 3,4-dichloro-l-phenylalanine, 2s-2-amino-3-3,4-dichlorophenyl propanoic acid, h-phe 3,4-dicl-oh, 3,4-dichloro-phenylalanine, l-3,4-dichlorophenlalanine, l-3,4-dichlorophe, h-3,4-dichloro-phe-oh, dichlorophe
PubChem CID 2761498
IUPAC Name (2S)-2-amino-3-(3,4-dichlorophenyl)propanoic acid
SMILES C1=CC(=C(C=C1CC(C(=O)O)N)Cl)Cl

Specifications

Quantity 1 g
Formula Weight 234.08
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3,4-Dichloro-L-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.