Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzaldoxime, 98%, Thermo Scientific™

Catalog No. AAL1020203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1020203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1020203 Supplier Thermo Scientific Chemicals Supplier No. L1020203

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5331-92-0
Molecular Formula C7H5Cl2NO
Molecular Weight (g/mol) 190.02
MDL Number MFCD00017592
InChI Key ROBIUDOANJUDHD-ONNFQVAWSA-N
Synonym 3,4-dichlorobenzaldoxime, 3,4-dichlorobenzaldehyde oxime, 1e-3,4-dichlorobenzaldehyde oxime, e-n-3,4-dichlorophenyl methylidene hydroxylamine, ne-n-3,4-dichlorophenyl methylidene hydroxylamine, 3,4-dichlorophenyl hydroxyimino methane, 3,4-dichlorobenzaldehyde oxime #, z-n-3,4-dichlorophenyl methylidene hydroxylamine
PubChem CID 5704835
IUPAC Name (NE)-N-[(3,4-dichlorophenyl)methylidene]hydroxylamine
SMILES O\N=C\C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Beilstein 2206792
Formula Weight 190.03
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorobenzaldoxime
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.