Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzoic Acid 99+%, Thermo Scientific™

Catalog No. AC163230250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC163230250 25 g
AC163231000 100 g
AC163235000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC163230250 Supplier Thermo Scientific Chemicals Supplier No. 163230250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-44-5
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002492
InChI Key VPHHJAOJUJHJKD-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609
PubChem CID 5817
ChEBI CHEBI:49401
IUPAC Name 3,4-dichlorobenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Linear Formula Cl2C6H3CO2H
Quantity 25 g
Beilstein 09, 343
Formula Weight 191.01
Chemical Name or Material 3, 4-Dichlorobenzoic acid, 99+%
EINECSNumber 200-099-2
RTECSNumber DG7175000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.