Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dichlorobenzoic acid-d3 | 350818-53-0 | 99.5% | 194.03 g·mol⁻1 | C7HD3Cl2O2 | 1 MG
SDP

Catalog No. 5000302453
Encompass_Preferred

3,4-dichlorobenzoic acid-d3 is the deuterium-labeled isotopologue of 3,4-dichlorobenzoic acid used as a stable-isotope tracer and internal standard in analytical chemistry. It is provided as a high-purity material suitable for quantitative workflows in NMR, GC-MS, and LC-MS.

  • High isotopic labeling for reliable tracing and quantitation.
  • Purity 99.5% by HPLC.
  • Molecular formula C7HD3Cl2O2; molecular weight 194.03 g·mol⁻1.
  • Supplied in small research pack sizes, including 1 mg.
  • Intended for research use only; not for human therapeutic use.

Catalog No. 50-003-02453 Supplier Medchemexpress LLC Supplier No. HYW015139S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-dichlorobenzoic acid-d3 is the deuterium-labeled isotopologue of 3,4-dichlorobenzoic acid used as a stable-isotope tracer and internal standard in analytical chemistry. It is provided as a high-purity material suitable for quantitative workflows in NMR, GC-MS, and LC-MS.

  • High isotopic labeling for reliable tracing and quantitation.
  • Purity 99.5% by HPLC.
  • Molecular formula C7HD3Cl2O2; molecular weight 194.03 g·mol⁻1.
  • Supplied in small research pack sizes, including 1 mg.
  • Intended for research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.