Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dichlorobenzoic acid-d3 | 350818-53-0 | 99.5% | 194.03 g/mol | C7HD3Cl2O2 | 10 MG
SDP

Catalog No. 5000303663
Encompass_Preferred

3,4-Dichlorobenzoic acid-d3 is the deuterium-labeled analogue of 3,4-dichlorobenzoic acid intended for use as an isotopic tracer or internal standard in analytical chemistry. It is provided with high isotopic and chemical purity for quantitative applications in NMR, GC-MS, and LC-MS.

  • High chemical purity (99.5%).
  • Deuterium-labeled for accurate mass spectrometry and NMR quantitation.
  • Suitable as an internal standard in GC-MS and LC-MS workflows.
  • Available in multiple small pack sizes for analytical use.
  • Molecular formula C7HD3Cl2O2 and molecular weight 194.03 g/mol.

Catalog No. 50-003-03663 Supplier Medchemexpress LLC Supplier No. HYW015139S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-Dichlorobenzoic acid-d3 is the deuterium-labeled analogue of 3,4-dichlorobenzoic acid intended for use as an isotopic tracer or internal standard in analytical chemistry. It is provided with high isotopic and chemical purity for quantitative applications in NMR, GC-MS, and LC-MS.

  • High chemical purity (99.5%).
  • Deuterium-labeled for accurate mass spectrometry and NMR quantitation.
  • Suitable as an internal standard in GC-MS and LC-MS workflows.
  • Available in multiple small pack sizes for analytical use.
  • Molecular formula C7HD3Cl2O2 and molecular weight 194.03 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.