Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dichlorobenzoic acid-d3 | 350818-53-0 | 99.5% | 194.03 g·mol⁻¹ | C7HD3Cl2O2 | 5 MG
SDP

Catalog No. 5000303371
Encompass_Preferred

3,4-Dichlorobenzoic acid-d3 is the deuterium-labeled analogue of 3,4-dichlorobenzoic acid (CAS 350818-53-0) supplied as a white to off-white solid high-purity isotope reagent for research use. It is intended for analytical applications including mass spectrometry internal standards, tracer experiments, and isotope-labeling investigations.

  • High isotopic purity (99.5%).
  • Solid, white to off-white form for convenient handling.
  • Molecular weight 194.03 g·mol⁻¹; formula C7HD3Cl2O2 for reference.
  • Suitable as an internal standard for mass spectrometry and quantitative assays.
  • Available in small milligram pack sizes for analytical studies.

Catalog No. 50-003-03371 Supplier Medchemexpress LLC Supplier No. HYW015139S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-Dichlorobenzoic acid-d3 is the deuterium-labeled analogue of 3,4-dichlorobenzoic acid (CAS 350818-53-0) supplied as a white to off-white solid high-purity isotope reagent for research use. It is intended for analytical applications including mass spectrometry internal standards, tracer experiments, and isotope-labeling investigations.

  • High isotopic purity (99.5%).
  • Solid, white to off-white form for convenient handling.
  • Molecular weight 194.03 g·mol⁻¹; formula C7HD3Cl2O2 for reference.
  • Suitable as an internal standard for mass spectrometry and quantitative assays.
  • Available in small milligram pack sizes for analytical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.