Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D034025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D034025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D034025G Supplier TCI America Supplier No. D034025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 51-44-5
Melting Point 205°C
Color White
Molecular Formula C7H4Cl2O2
Quantity 25 g
MDL Number MFCD00002492
Synonym benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609
InChI Key VPHHJAOJUJHJKD-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=C(Cl)C(Cl)=C1
IUPAC Name 3,4-dichlorobenzoic acid
Molecular Weight (g/mol) 191.01
PubChem CID 5817
ChEBI CHEBI:49401
Formula Weight 191.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorobenzoic Acid
Show More Show Less
EINECSNumber (3)-1410&(3)-1424
RTECSNumber DG7175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.