Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. D341525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D341525G 25 g
D34155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D341525G Supplier TCI America Supplier No. D341525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6574-99-8
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00016379
InChI Key KUWBYWUSERRVQP-UHFFFAOYSA-N
Synonym benzonitrile, 3,4-dichloro, 3,4-dichloro benzonitrile, 3.4-dichlorobenzonitrile, 3, 4-dichlorobenzonitrile, 3,4-dichlorobenzenecarbonitrile, 3,4-dichloro-benzonitrile, pubchem3688, acmc-1b9ly, 3,4-dichlorobenzonitrile, buttpark 87\02-72
PubChem CID 81051
IUPAC Name 3,4-dichlorobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Cl)Cl

Specifications

Melting Point 73°C
Color White
Quantity 25 g
UN Number 3439
Formula Weight 172.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.