Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzyl Alcohol, MP Biomedicals

Catalog No. ICN227402 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227402 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227402 Supplier MP Biomedicals, Inc Supplier No. 0522740205

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1805-32-9
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
InChI Key FVJIUQSKXOYFKG-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl alcohol, 3,4-dichlorophenyl methanol, benzenemethanol, 3,4-dichloro, 3,4-dichlorobenzylalcohol, rarechem al bd 0536, benzyl alcohol, 3,4-dichloro, labotest-bb lt02003790, benzenemethanol,4-dichloro, benzyl alcohol,4-dichloro, 3,4-dichloro-1-hydroxymethyl-benzene
PubChem CID 15728
IUPAC Name (3,4-dichlorophenyl)methanol
SMILES C1=CC(=C(C=C1CO)Cl)Cl

Specifications

Quantity 10 g
Formula Weight 177.03
Chemical Name or Material 3, 4-Dichlorobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.