Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzyl bromide, 97%

Catalog No. p-3736167
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC326160250 25 g Glass bottle
AC326161000 100 g Glass bottle
AC326165000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC326160250 Supplier Thermo Scientific Chemicals Supplier No. 326160250
Only null left
Add to Cart

CAS: 18880-04-1 | C7H5BrCl2 | 239.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18880-04-1
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.93
MDL Number MFCD00041363
InChI Key XLWSBDFQAJXCQX-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl bromide, 4-bromomethyl-1,2-dichlorobenzene, benzene, 4-bromomethyl-1,2-dichloro, 3,4-dichlorobenzylbromide, alpha-bromo-3,4-dichlorotoluene, acmc-209erq, 3,4 dichlorobenzyl bromide, 3,4-dichloro-benzyl bromide, ksc493a6b, timtec-bb sbb005763
PubChem CID 639726
IUPAC Name 4-(bromomethyl)-1,2-dichlorobenzene
SMILES C1=CC(=C(C=C1CBr)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.6500g/mL
Boiling Point 115°C to 120°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6050 to 1.607
Quantity 25 g
Specific Gravity 1.65
Formula Weight 239.93
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichlorobenzyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.