Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dichlorobenzyl chloride

Catalog No. 259179100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-917-9100G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-917-9100G Supplier Sigma Aldrich Supplier No. 259179100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,4-Dichlorobenzyl chloride

Specifications

CAS 102-47-6
Density 1.411 g/mL (at 25°C (literature))
Boiling Point 122°C to 124°C (14 mmHg)
Molecular Formula C7H5Cl3
Refractive Index n20/D 1.577 (literature)
Linear Formula Cl2C6H3CH2Cl
MDL Number MFCD00000906
Quantity 100 mg
Synonym alpha,3,4-Trichlorotoluene
Molecular Weight (g/mol) 195.47
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.