Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. T040125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T040125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T040125G Supplier TCI America Supplier No. T040125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 102-47-6
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
MDL Number MFCD00000906
InChI Key YZIFVWOCPGPNHB-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl chloride, 1,2-dichloro-4-chloromethyl benzene, alpha,3,4-trichlorotoluene, benzene, 1,2-dichloro-4-chloromethyl, unii-451pb8smie, 1,2-dichloro-4-chloromethyl-benzene, 3,4-dichlorobenzylchloride, 451pb8smie, 3,4-dichlorophenylmethyl chloride, toluene, .alpha.,3,4-trichloro
PubChem CID 7609
IUPAC Name 1,2-dichloro-4-(chloromethyl)benzene
SMILES C1=CC(=C(C=C1CCl)Cl)Cl

Specifications

Color Colorless
Boiling Point 241°C
Quantity 25 g
UN Number 3265
Formula Weight 195.47
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dichlorobenzyl Chloride
EINECSNumber (3)-0091
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.