Learn More
Apexbio Technology LLC 3,4-Dichloroisocoumarin 51050-59-0 10mg
SDP
Catalog No. 5000096023
Encompass_Preferred
3 4-Dichloroisocoumarin (CAS 51050-59-0) is a chlorinated derivative of isocoumarin that functions as an inhibitor of serine proteases This compound interacts with the active site serine residue of the enzyme thereby suppressing proteolytic activity Serine proteases play essential roles in various physiological processes including digestion immune response and cell signaling pathways 3 4-Dichloroisocoumarin is widely utilized as a biochemical tool for investigating serine protease function and enzymatic mechanisms in molecular and cellular studies
Catalog No. 50-000-96023 Supplier Apexbio Technology LLC Supplier No. A993510
3 4-Dichloroisocoumarin (CAS 51050-59-0) is a chlorinated derivative of isocoumarin that functions as an inhibitor of serine proteases This compound interacts with the active site serine residue of the enzyme thereby suppressing proteolytic activity Serine proteases play essential roles in various physiological processes including digestion immune response and cell signaling pathways 3 4-Dichloroisocoumarin is widely utilized as a biochemical tool for investigating serine protease function and enzymatic mechanisms in molecular and cellular studies