Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4'-Dichloropropiophenone, 98%

Catalog No. AAA1874006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1874006 5 g
AAA1874014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1874006 Supplier Thermo Scientific Chemicals Supplier No. A1874006
Add to Cart

CAS: 6582-42-9 | C9H8Cl2O | 203.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6582-42-9
Molecular Formula C9H8Cl2O
Molecular Weight (g/mol) 203.06
MDL Number MFCD00000554
InChI Key AJUAASWQUWIMHM-UHFFFAOYSA-N
Synonym 3',4'-dichloropropiophenone, 1-3,4-dichlorophenyl propan-1-one, 1-3,4-dichlorophenyl-1-propanone, 1-propanone, 1-3,4-dichlorophenyl, 1-propanone,1-3,4-dichlorophenyl, enamine_003892, acmc-1b5az, 3,4-dichloro propiophenone, 1-3,4-dichlorophenyl-1-propanone #
PubChem CID 599598
IUPAC Name 1-(3,4-dichlorophenyl)propan-1-one
SMILES CC(=O)CC1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 44°C to 46°C
Boiling Point 136°C to 140°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 203.07
Percent Purity 98%
Chemical Name or Material 3',4'-Dichloropropiophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-511-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.