Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4'-Dichloropropiophenone 98.0+%, TCI America™
SDP

Catalog No. D26135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26135G 5 g
D261325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26135G Supplier TCI America Supplier No. D26135G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3946-29-0
Molecular Formula C9H8Cl2O
Molecular Weight (g/mol) 203.06
MDL Number MFCD00000992
InChI Key AYFJBHFMQODYBC-UHFFFAOYSA-N
Synonym 3,4'-dichloropropiophenone, 3-chloro-1-4-chlorophenyl propan-1-one, 1-propanone, 3-chloro-1-4-chlorophenyl, 2-chloroethyl-4-chlorophenylketone, pubchem13270, acmc-209j5n, beta,4-dichloropropiophenone, 4',beta-dichloropropiophenone, 3-chloro-4'-chloropropiophenone, attercop-chm at130375
PubChem CID 77550
IUPAC Name 3-chloro-1-(4-chlorophenyl)propan-1-one
SMILES ClCCC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 51°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 203.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4′-Dichloropropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.