Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diethoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. D045125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D045125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D045125G Supplier TCI America Supplier No. D045125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2029-94-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00016607
InChI Key SSTRYEXQYQGGAS-UHFFFAOYSA-N
PubChem CID 74847
IUPAC Name 3,4-diethoxybenzaldehyde
SMILES CCOC1=C(C=C(C=C1)C=O)OCC

Specifications

Color White-Yellow
Boiling Point 165°C
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,4-Diethoxybenzaldehyde
EINECSNumber (3)-2665
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.