Learn More
Medchemexpress LLC 3-[4-(diethylamino)-3-nitrophenyl]-4-methyl-4,5-dihydro-1H-pyridazin-6-one | 328104-79-6 | MFCD01829240 | 99.5% | 304.34 g/mol | C15H20N4O3 | 10 MG
SDP
Catalog No. 5000301938
Encompass_Preferred
DNMDP is a research small molecule inhibitor of phosphodiesterase 3A (PDE3A) that promotes formation of a PDE3A-Schlafen 12 (SLFN12) complex, producing selective cytotoxicity in susceptible cancer cell lines. It is supplied as a powder for experimental in vitro and in vivo use.
Catalog No. 50-003-01938 Supplier Medchemexpress LLC Supplier No. HYW02869010MG
DNMDP is a research small molecule inhibitor of phosphodiesterase 3A (PDE3A) that promotes formation of a PDE3A-Schlafen 12 (SLFN12) complex, producing selective cytotoxicity in susceptible cancer cell lines. It is supplied as a powder for experimental in vitro and in vivo use.