Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluoro-2,5-bis(trimethylsilyl)thiophene 95.0+%, TCI America™
SDP

Catalog No. D37011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37011G Supplier TCI America Supplier No. D37011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 347838-12-4
Molecular Formula C10H18F2SSi2
Molecular Weight (g/mol) 264.48
MDL Number MFCD11036246
InChI Key BIIDYHMAKPKADL-UHFFFAOYSA-N
PubChem CID 44629942
IUPAC Name [3,4-difluoro-5-(trimethylsilyl)thiophen-2-yl]trimethylsilane
SMILES C[Si](C)(C)C1=C(F)C(F)=C(S1)[Si](C)(C)C

Specifications

Boiling Point 98°C
Quantity 1 g
Formula Weight 264.48
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluoro-2,5-bis(trimethylsilyl)thiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.