Learn More
Indofine Chemical 3,4-Difluoroanili, 98%, 100 Gm
SDP
Supplier:  Indofine Chemical 21459 100 GM
Encompass_Preferred
3,4-DIFLUOROANILINE 100 GM, Fluoro Organics, C6H5F2N, MW: 129.11, 0.98, 3863-11-4, MFCD00007761
Catalog No. 50-913-718