Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluorobenzenethiol 97.0+%, TCI America™
SDP

Catalog No. D41215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41215G 5 g
D412125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41215G Supplier TCI America Supplier No. D41215G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60811-24-7
Molecular Formula C6H3F2S
Molecular Weight (g/mol) 145.15
MDL Number MFCD03094279
InChI Key BGVRHDQMTMPAEZ-UHFFFAOYSA-M
Synonym 3,4-difluorothiophenol, 3,4-difluoro thiophenol, 3,4-difluorobenzene-1-thiol, benzenethiol, 3,4-difluoro, pubchem6772, 3,4-difluoro-thiophenol, 3.4-difluorobenzenethiol, acmc-1b3bf, 3,4-difluoro benzenethiol, 3,4-difluorobenzene-thiol
PubChem CID 2778378
IUPAC Name (3,4-difluorophenyl)sulfanide
SMILES FC1=CC=C([S-])C=C1F

Specifications

Color Yellow
Boiling Point 70°C
Quantity 5 g
UN Number 3336
Formula Weight 146.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorobenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.