Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluorobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D24155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D24155G Supplier TCI America Supplier No. D24155G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85118-05-4
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD00010628
InChI Key GNQLTCVBSGVGHC-UHFFFAOYSA-N
Synonym 3,4-difluorobenzyl alcohol, 3,4-difluorophenyl methanol, benzenemethanol, 3,4-difluoro, 3,4-difluorobenzylalcohol, 3,4-difluoro-phenyl-methanol, 3,4-difluorophenyl methan-1-ol, 3, 4-difluorobenzyl alcohol, pubchem4916, acmc-209q4o, ksc493q4f
PubChem CID 522833
IUPAC Name (3,4-difluorophenyl)methanol
SMILES OCC1=CC=C(F)C(F)=C1

Specifications

Quantity 5 g
Formula Weight 144.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.