Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluoronitrobenzene, 99%

Catalog No. AC241931000
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC241931000 100 mL Glass bottle
AC241930250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC241931000 Supplier Thermo Scientific Chemicals Supplier No. 241931000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 369-34-6 | C6H3F2NO2 | 159.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-34-6
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00007198
InChI Key RUBQQRMAWLSCCJ-UHFFFAOYSA-N
Synonym 3,4-difluoronitrobenzene, benzene, 1,2-difluoro-4-nitro, 1-nitro-3,4-difluorobenzene, 1,2-difluoro-4-nitro-benzene, 3,4-difluoro nitrobenzene, pubchem2290, 3,4-difluornitrobenzene, 3.4-difluoronitrobenzene, 3, 4-difluoronitrobenzene, 3,4-difluoro-nitrobenzene
PubChem CID 123053
IUPAC Name 1,2-difluoro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.4410g/mL
Boiling Point 80°C to 81°C (14.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5080 to 1.51
Linear Formula F2(C6H3)NO2
Quantity 100 mL
Beilstein 05, III, 609
Specific Gravity 1.441
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 159.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3, 4-Difluoronitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-718-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.