Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluorophenylacetic acid, 98%, Thermo Scientific™

Catalog No. AC311640050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC311640050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC311640050 Supplier Thermo Scientific Chemicals Supplier No. 311640050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 658-93-5
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00010002
InChI Key YCAKYFIYUHHCKW-UHFFFAOYSA-N
Synonym 3,4-difluorophenylacetic acid, 2-3,4-difluorophenyl acetic acid, 3,4-difluorophenyl acetic acid, benzeneacetic acid, 3,4-difluoro, 3,4-difluorobenzeneacetic acid, 3,4-difluorophenylaceticacid, pubchem4154, acmc-209nst, ksc355a2l, rarechem al bo 0250
PubChem CID 69571
IUPAC Name 2-(3,4-difluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 47°C to 51°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula F2F6H3CH2COOH
Quantity 5 g
Formula Weight 172.13
Percent Purity ≥97.5%
Physical Form Crystalline Needles
Chemical Name or Material 3, 4-Difluorophenylacetic acid
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.