Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Difluorophenylmagnesium Bromide Solution

Catalog No. 56103750ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-103-750ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 56-103-750ML Supplier Sigma Aldrich Supplier No. 56103750ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,4-Difluorophenylmagnesium bromide can be used:To prepare 3,4-difluorophenylboronic acid, a key building block employed in the synthesis of norepinephrine transporter selective molecules.In the synthesis of trimethoxybenzophenone derived tubulin polymerization inhibitors.In one of the key synthetic steps for the preparation of diarylketoxime derivatives as potent MCH-1R antagonists.In the synthesis of biologically important hydroxybupropion analogs useful in smoking cessation.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 89-54-3
Melting Point 184°C to 188°C
Molecular Formula C7H6ClNO2
Linear Formula H2NC6H3(Cl)CO2H
MDL Number MFCD00007845
Quantity 50 mL
Molecular Weight (g/mol) 171.58 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.