Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4'-Difluoropropiophenone, 97%

Catalog No. AAA1725706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1725706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1725706 Supplier Thermo Scientific Chemicals Supplier No. A1725706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23384-72-7 | C9H8F2O | 170.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23384-72-7
Molecular Formula C9H8F2O
Molecular Weight (g/mol) 170.16
MDL Number MFCD00015505
InChI Key FSZOBSMJJFHVCQ-UHFFFAOYSA-N
Synonym 1-3,4-difluorophenyl propan-1-one, 3',4'-difluoropropiophenone, 3,4-difluoropropiophenone, propiophenone, 3',4'-difluoro, 1-propanone, 1-3,4-difluorophenyl, acmc-1cgp0, ksc551g8r, 1-3,4-difluorophenyl-1-propanone, 1-propanone,1-3,4-difluorophenyl, 1-3,4-difluorophenyl-1-propanone #
PubChem CID 520050
IUPAC Name 1-(3,4-difluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(F)C(F)=C1

Specifications

Boiling Point 64°C to 65°C (1.8 mmHg)
Refractive Index 1.488
Quantity 5 g
Beilstein 2088650
Formula Weight 170.16
Percent Purity 97%
Chemical Name or Material 3',4'-Difluoropropiophenone
EINECSNumber 245-627-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.