Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dihydro-2H-Pyran-2-Methanol

Catalog No. 5566525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-665-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 55-665-25ML Supplier Sigma Aldrich Supplier No. 5566525ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Preparation of supported dihydropyran for mild immobilization and cleavage of alcohols

Specifications

CAS 20780-54-5
Density 1.051 g/mL at 25°C
Boiling Point 192°C to 194°C
Molecular Formula C8H8O
Linear Formula C8H8O
Refractive Index n20/D 1.478; n20/D 1.479
Quantity 25 mL
MDL Number MFCD00064310
Synonym 2-Hydroxymethyl-3,4-dihydro-2H-pyran; 3,4-Dihydro-2H-pyran-2-ylmethanol
Molecular Weight (g/mol) 120.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.