Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2H-pyran-2-methanol 97.0+%, TCI America™
SDP

Catalog No. D32951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32951G 1 g
D32955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32951G Supplier TCI America Supplier No. D32951G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3749-36-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
InChI Key XMICBFRKICBBKD-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-pyran-2-methanol, 2-hydroxymethyl-3,4-dihydro-2h-pyran, dhp linker, 3,4-dihydro-2h-pyran-2-yl methanol, 2h-pyran-2-methanol, 3,4-dihydro, dhp resin, ellmans dhp resin, zlchem 376, pubchem9703, acmc-209itf
PubChem CID 95559
IUPAC Name 3,4-dihydro-2H-pyran-2-ylmethanol
SMILES C1CC(OC=C1)CO

Specifications

Color Colorless
Boiling Point 93°C
Quantity 1 g
Formula Weight 114.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dihydro-2H-pyran-2-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.