Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dihydro-6,7-isoquinolinediol | 4602-83-9 | MFCD00143490 | 98.7% | 163.17 g/mol | C9H9NO2 | 10 MG
SDP

Catalog No. 5000281841
Encompass_Preferred

3,4-dihydro-6,7-isoquinolinediol is a small-molecule research reagent commonly used in biochemical and pharmacological studies where isoquinoline derivatives are required. The compound is supplied as a high-purity solid suitable for laboratory experiments and analytical work.

  • High purity: 98.7%.
  • Molecular formula: C9H9NO2.
  • Molecular weight: 163.17 g/mol.
  • CAS number: 4602-83-9.
  • Available pack sizes include 10 mg, 25 mg, 50 mg, and 100 mg.

Catalog No. 50-002-81841 Supplier Medchemexpress LLC Supplier No. HYW34154710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-dihydro-6,7-isoquinolinediol is a small-molecule research reagent commonly used in biochemical and pharmacological studies where isoquinoline derivatives are required. The compound is supplied as a high-purity solid suitable for laboratory experiments and analytical work.

  • High purity: 98.7%.
  • Molecular formula: C9H9NO2.
  • Molecular weight: 163.17 g/mol.
  • CAS number: 4602-83-9.
  • Available pack sizes include 10 mg, 25 mg, 50 mg, and 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.