Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dihydro-6,7-isoquinolinediol | 4602-83-9 | MFCD00143490 | 98.7% | 163.17 g/mol | C9H9NO2 | 100 MG
SDP

Catalog No. 5000282551
Encompass_Preferred

3,4-Dihydro-6,7-isoquinolinediol is a small-molecule research compound used in biochemical and pharmacological studies and reported to act as a β2-adrenergic receptor agonist with measurable cellular activity. It is supplied as a solid in milligram-scale vials for laboratory use and includes recommended storage and solvent stability conditions for research handling.

  • Molecular formula: C9H9NO2.
  • Molecular weight: 163.17 g/mol.
  • Cas number: 4602-83-9.
  • Purity: 98.7% (reported).
  • Available package sizes: 10 MG, 25 MG, 50 MG, 100 MG, 200 MG, 500 MG.
  • Storage: store at 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month) when under nitrogen.
  • Intended use: for biochemical and pharmacological research only, not for human use.

Catalog No. 50-002-82551 Supplier Medchemexpress LLC Supplier No. HYW341547100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-Dihydro-6,7-isoquinolinediol is a small-molecule research compound used in biochemical and pharmacological studies and reported to act as a β2-adrenergic receptor agonist with measurable cellular activity. It is supplied as a solid in milligram-scale vials for laboratory use and includes recommended storage and solvent stability conditions for research handling.

  • Molecular formula: C9H9NO2.
  • Molecular weight: 163.17 g/mol.
  • Cas number: 4602-83-9.
  • Purity: 98.7% (reported).
  • Available package sizes: 10 MG, 25 MG, 50 MG, 100 MG, 200 MG, 500 MG.
  • Storage: store at 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month) when under nitrogen.
  • Intended use: for biochemical and pharmacological research only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.