Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dihydro-6,7-isoquinolinediol | 4602-83-9 | MFCD00143490 | 98.7% | 163.17 g·mol⁻¹ | C9H9NO2 | 50 MG
SDP

Catalog No. 5000282100
Encompass_Preferred

3,4-Dihydro-6,7-isoquinolinediol is a small-molecule research compound (C9H9NO2) used in analytical and biochemical studies. It is supplied with high reported purity and has associated analytical and safety documentation for laboratory handling and storage.

  • High purity: 98.7%.
  • Molecular weight: 163.17 g·mol⁻¹.
  • Chemical formula: C9H9NO2.
  • CAS number: 4602-83-9.
  • Available in milligram quantities (e.g., 10 mg-100 mg) for research use.
  • Storage recommendations: store at 4°C under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-82100 Supplier Medchemexpress LLC Supplier No. HYW34154750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-Dihydro-6,7-isoquinolinediol is a small-molecule research compound (C9H9NO2) used in analytical and biochemical studies. It is supplied with high reported purity and has associated analytical and safety documentation for laboratory handling and storage.

  • High purity: 98.7%.
  • Molecular weight: 163.17 g·mol⁻¹.
  • Chemical formula: C9H9NO2.
  • CAS number: 4602-83-9.
  • Available in milligram quantities (e.g., 10 mg-100 mg) for research use.
  • Storage recommendations: store at 4°C under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.