Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-Dihydro-6-methoxy-2,2-dimethyl-2H-naphtho[1,2-b]pyran | 20213-26-7 | 1 MG
SDP

Catalog No. 5000418637
Encompass_Preferred

Dihydrolapachenole is a chemical compound with a molecular formula of C16H18O2 and a molecular weight of 242.31. It typically appears as a white to off-white solid and has a high purity of 98.57% as determined by HPLC. This product is designed for research applications and has not been validated for medical use.

  • Chemical name: 3,4-Dihydro-6-methoxy-2,2-dimethyl-2H-naphtho[1,2-b]pyran
  • Molecular formula: C16H18O2
  • Molecular weight: 242.31
  • Appearance: White to off-white solid
  • Purity (HPLC): 98.57%
  • Store at -20°C, sealed, away from moisture and light
  • For research use only

Catalog No. 50-004-18637 Supplier Medchemexpress LLC Supplier No. HYN76381MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydrolapachenole is a chemical compound with a molecular formula of C16H18O2 and a molecular weight of 242.31. It typically appears as a white to off-white solid and has a high purity of 98.57% as determined by HPLC. This product is designed for research applications and has not been validated for medical use.

  • Chemical name: 3,4-Dihydro-6-methoxy-2,2-dimethyl-2H-naphtho[1,2-b]pyran
  • Molecular formula: C16H18O2
  • Molecular weight: 242.31
  • Appearance: White to off-white solid
  • Purity (HPLC): 98.57%
  • Store at -20°C, sealed, away from moisture and light
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.