Learn More
Cayman Chemical 3,4-dihydro Naratriptan, Purity: ≥95%, CAS: 121679-20-7, 10 mg
SDP
Catalog No. 502966975
Encompass_Preferred
A process impurity formed during the preparation of naratriptan
Catalog No. 50-296-6975 Supplier Cayman Chemical Supplier No. 1789010 MG
A process impurity formed during the preparation of naratriptan