Learn More
Cayman Chemical 3,4-Dihydroquinolin-2(1H)-one, Purity: ≥98%, CAS: 553-03-7, 5 g
SDP
Catalog No. 502992243
Encompass_Preferred
A building block; has been used in the synthesis of compounds with antidepressant and anticonvulsant activities, as well as dual ligands of σ receptors and GluN2b that have antioxidant and neuroprotective properties
Catalog No. 50-299-2243 Supplier Cayman Chemical Supplier No. 317015 G
A building block; has been used in the synthesis of compounds with antidepressant and anticonvulsant activities, as well as dual ligands of σ receptors and GluN2b that have antioxidant and neuroprotective properties