Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dihydroxy-1-butene

Catalog No. 488216100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-821-6100ML 100 mL
48-821-625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48-821-6100ML Supplier Sigma Aldrich Supplier No. 488216100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,4-dihydroxy-1-butene, also known as 3-butene-1,2-diol (bddiol), is a metabolite of 1,3-butadiene. it forms the precursor for synthesizing different chiral building blocks. bddiol can undergo oxidation to form hydroxymethylvinyl ketone (hmvk). 1,2-epoxy-3-butene (eb) on hydrolysis in the presence of epoxide hydrolases (eh) forms bddiol.3,4-dihydroxy-1-butene can be used:
  • As a reactant to synthesize cyclic organic carbonates by continuous flow procedure
  • To prepare substituted oxazolidinone ligands used to target medicinally relevant rnas

Specifications

CAS 497-06-3
Density 1.047 g/mL (at 25°C (literature))
Boiling Point 195°C (733 mmHg)
Molecular Formula C4H8O2
Linear Formula CH2=CHCH(OH)CH2OH
MDL Number MFCD00239410
Quantity 100 mL
Synonym 3-Butene-1,2-diol
Molecular Weight (g/mol) 88.11
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.