Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
3,4-Dihydroxy-3-cyclobutene-1,2-dione
Spécifications
Spécifications
| CAS | 2892-51-5 |
| Melting Point | >300°C |
| Molecular Formula | C4H2O4 |
| Linear Formula | (HO)2C4(=O)2 |
| MDL Number | MFCD00001334 |
| Quantity | 25 g |
| Synonym | Squaric acid |
| Molecular Weight (g/mol) | 114.06 |
| Percent Purity | ≥99.0% (HPLC) |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'