Learn More
Cayman Chemical 3 4-DIhydroxybenzaldehyde 100g
SDP
Catalog No. 502937907
Encompass_Preferred
A polyphenol with diverse biological activities; active against MRSA at 0.01 UG/ml; prevents hexavalent chromium-induced formation of ROS and NOS in a concentration-dependent manner and increases GSH in isolated human erythrocytes; selectively inhibits human DNA topoisomerase II (IC50 = 150 µM) over human topoisomerase I and a variety of mammalian polymerases (IC50s = >200 µM); inhibits proliferation of HT-29 cells at 362 µM but not HCT116 cells at 100 µM
Catalog No. 50-293-7907 Supplier Cayman Chemical Supplier No. 27547100 G
A polyphenol with diverse biological activities; active against MRSA at 0.01 UG/ml; prevents hexavalent chromium-induced formation of ROS and NOS in a concentration-dependent manner and increases GSH in isolated human erythrocytes; selectively inhibits human DNA topoisomerase II (IC50 = 150 µM) over human topoisomerase I and a variety of mammalian polymerases (IC50s = >200 µM); inhibits proliferation of HT-29 cells at 362 µM but not HCT116 cells at 100 µM