Learn More
Apexbio Technology LLC 3,4-Dihydroxybenzaldehyde 139-85-5 20mg
SDP
Catalog No. 5000093035
Encompass_Preferred
3 4-Dihydroxybenzaldehyde (139-85-5) is a small-molecule compound known for its antioxidative and antimicrobial activities It exerts its biological effects primarily by neutralizing reactive oxygen species (ROS) and suppressing microbial growth via interactions with cellular membranes and metabolic pathways In in vitro assays 3 4-Dihydroxybenzaldehyde demonstrates inhibitory activity against various bacterial strains and inflammatory mediators with reported IC50 values ranging from approximately 10 to 50 M depending on the cell type and experimental conditions Based on these pharmacological properties 3 4-Dihydroxybenzaldehyde holds research potential in studies of oxidative stress inflammatory responses bacterial infection models drug-resistance mechanisms and therapeutic investigations in cell-based and molecular biology assays
Catalog No. 50-000-93035 Supplier Apexbio Technology LLC Supplier No. N227620
3 4-Dihydroxybenzaldehyde (139-85-5) is a small-molecule compound known for its antioxidative and antimicrobial activities It exerts its biological effects primarily by neutralizing reactive oxygen species (ROS) and suppressing microbial growth via interactions with cellular membranes and metabolic pathways In in vitro assays 3 4-Dihydroxybenzaldehyde demonstrates inhibitory activity against various bacterial strains and inflammatory mediators with reported IC50 values ranging from approximately 10 to 50 M depending on the cell type and experimental conditions Based on these pharmacological properties 3 4-Dihydroxybenzaldehyde holds research potential in studies of oxidative stress inflammatory responses bacterial infection models drug-resistance mechanisms and therapeutic investigations in cell-based and molecular biology assays