Learn More
Selleck Chemical LLC 3.4-Dihydroxybenzaldehyde S3952-25mg
SDP
Catalog No. 503110080
Encompass_Preferred
3 4-Dihydroxybenzaldehyde (Protocatechuic aldehyde) is a phenolic aldehyde a precursor in vanillin synthesis via biotransformation by cell cultures of Capsicum frutescens a type of chili pepper
Catalog No. 50-311-0080 Supplier Selleck Chemical LLC Supplier No. S395225MG
3 4-Dihydroxybenzaldehyde (Protocatechuic aldehyde) is a phenolic aldehyde a precursor in vanillin synthesis via biotransformation by cell cultures of Capsicum frutescens a type of chili pepper