Learn More
Cayman Chemical 3,4-Dihydroxybenzaldehyde, Purity: ≥98%, CAS: 139-85-5, 25 g
SDP
Catalog No. 502941625
Encompass_Preferred
A polyphenol with diverse biological activities; active against MRSA at 0.01 µg/ml; prevents hexavalent chromium-induced formation of ROS and NOS in a concentration-dependent manner and increases GSH in isolated human erythrocytes; selectively inhibits human DNA topoisomerase II (IC50 = 150 µM) over human topoisomerase I and a variety of mammalian polymerases (IC50s = >200 µM); inhibits proliferation of HT-29 cells at 362 µM but not HCT116 cells at 100 µM
Catalog No. 50-294-1625 Supplier Cayman Chemical Supplier No. 2754725 G
A polyphenol with diverse biological activities; active against MRSA at 0.01 µg/ml; prevents hexavalent chromium-induced formation of ROS and NOS in a concentration-dependent manner and increases GSH in isolated human erythrocytes; selectively inhibits human DNA topoisomerase II (IC50 = 150 µM) over human topoisomerase I and a variety of mammalian polymerases (IC50s = >200 µM); inhibits proliferation of HT-29 cells at 362 µM but not HCT116 cells at 100 µM