Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dihydroxybenzhydrazide, 97%

Click to view available options
Quantity:
5g
25g
39635-11-5
C7H8N2O3
168.152
MFCD00017064
WGXWEXNJRZMIPT-UHFFFAOYSA-N
3,4-dihydroxybenzhydrazide,3,4-dihydroxy-benzoic acid hydrazide,4-???benzene-1,2-diol,protocatechuhydrazide,acmc-1ad2t,3,4-dihydroxy-benzohydrazide,3,4-bis oxidanyl benzohydrazide,3,4-dihydroxybenzoic acid hydrazide,3,4-dihydroxy-benzoesaeure-hydrazid,3,4-dihydroxybenzoic acid, hydrazide
586300
3,4-dihydroxybenzohydrazide
C1=CC(=C(C=C1C(=O)NN)O)O
39635-11-5
C7H8N2O3
168.152
MFCD00017064
WGXWEXNJRZMIPT-UHFFFAOYSA-N
3,4-dihydroxybenzhydrazide,3,4-dihydroxy-benzoic acid hydrazide,4-???benzene-1,2-diol,protocatechuhydrazide,acmc-1ad2t,3,4-dihydroxy-benzohydrazide,3,4-bis oxidanyl benzohydrazide,3,4-dihydroxybenzoic acid hydrazide,3,4-dihydroxy-benzoesaeure-hydrazid,3,4-dihydroxybenzoic acid, hydrazide
586300
3,4-dihydroxybenzohydrazide
C1=CC(=C(C=C1C(=O)NN)O)O
Quantity 5g
Melting Point ~265°C (decomposition)
Beilstein 2642968
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 3,4-Dihydroxybenzhydrazide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning