Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dihydroxybenzylamine hydrobromide

Catalog No. 858781250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-878-1250MG 250 mg
85-878-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 85-878-1250MG Supplier Sigma Aldrich Supplier No. 858781250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,4-Dihydroxybenzylamine hydrobromide

Specifications

CAS 16290-26-9
Melting Point 184°C to 186°C (lit.)
Molecular Formula C7H9NO2 -+ HBr
Linear Formula (HO)2C6H3CH2NH2 -+ HBr
MDL Number MFCD00012859
Quantity 250 mg
Synonym 4-(Aminomethyl)catechol hydrobromide; DHBA hydrobromide
Molecular Weight (g/mol) 220.06
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.