Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydroxybenzylamine Hydrobromide, MP Biomedicals™

Catalog No. ICN15374083 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15374083 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15374083 Supplier MP Biomedicals, Inc Supplier No. 0215374083
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16290-26-9
Molecular Formula C7H10BrNO2
Molecular Weight (g/mol) 220.07
MDL Number MFCD00012859
InChI Key BVFZTXFCZAXSHN-UHFFFAOYSA-N
Synonym 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, dhba hydrobromide, acmc-1afa4, 1, 4-aminomethyl-, hydrobromide, 4-aminomethyl catechol hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide
PubChem CID 13343562
IUPAC Name hydrogen 4-(aminomethyl)benzene-1,2-diol bromide
SMILES [H+].[Br-].NCC1=CC=C(O)C(O)=C1

Specifications

Quantity 250 mg
Formula Weight 220.1
Physical Form Solid
Chemical Name or Material 3, 4-Dihydroxybenzylamine Hydrobromide
storageNote1 Protect from light.
Recommended Storage Store at -20°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.