Learn More
Cayman Chemical 3',4'-Dihydroxyflavone, Purity: ≥95%, CAS: 4143-64-0, 50 mg
SDP
Catalog No. 502910410
Encompass_Preferred
A synthetic flavonoid with diverse biological activities; inhibits TNKS1 and TNKS2 (IC50s = 0.23 and 0.17 µM for the human enzymes, respectively); active against L. donovani, T. brucei, and T. cruzi (IC50s = 2, 1.1, and 10.1 µg/ml, respectively); induces apoptosis in and inhibits migration of GL15 cells at 50 µM; prevents LPS-induced increases in brain levels of COX-2 and iNOS in a mouse model of neuroinflammation at 5 mg/kg
Catalog No. 50-291-0410 Supplier Cayman Chemical Supplier No. 3522650 MG
A synthetic flavonoid with diverse biological activities; inhibits TNKS1 and TNKS2 (IC50s = 0.23 and 0.17 µM for the human enzymes, respectively); active against L. donovani, T. brucei, and T. cruzi (IC50s = 2, 1.1, and 10.1 µg/ml, respectively); induces apoptosis in and inhibits migration of GL15 cells at 50 µM; prevents LPS-induced increases in brain levels of COX-2 and iNOS in a mouse model of neuroinflammation at 5 mg/kg