Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Dihydroxyhydrocinnamic acid

Catalog No. 501855499
Encompass
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-5499 2.5 g
10-260-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-5499 Supplier Sigma Aldrich Supplier No. 1026012.5G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,4-Dihydroxyhydrocinnamic acid
TRUSTED_SUSTAINABILITY

Specifications

CAS 1078-61-1
Melting Point 136°C to 139°C
Molecular Formula C9H10O4
Linear Formula (HO)2C6H3CH2CH2CO2H
Quantity 2.5 g
MDL Number MFCD00002776
Synonym 3-(3,4-Dihydroxyphenyl)propionic acid; Hydrocaffeic acid
Molecular Weight (g/mol) 182.17
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.