Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-Dihydroxymandelic acid | 775-01-9 | 98.0% | 184.15 | 10 MG
SDP

Catalog No. 5000437646
Encompass_Preferred

3,4-Dihydroxymandelic acid is an endogenous metabolite of norepinephrine. It is categorized as a human endogenous metabolite, belonging to the structural classifications of phenols, polyphenols, ketones, aldehydes, and acids. For research use only, it is produced by commensal microbiota and has been shown to promote chemotaxis and virulence gene expression in Enterohemorrhagic Escherichia coli.

  • Endogenous metabolite of norepinephrine
  • High purity, 98.0%
  • Molecular weight of 184.15
  • Chemical formula C8H8O5
  • Appears as a white to off-white solid
  • Soluble in water and DMSO

Catalog No. 50-004-37646 Supplier Medchemexpress LLC Supplier No. HY11347410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-Dihydroxymandelic acid is an endogenous metabolite of norepinephrine. It is categorized as a human endogenous metabolite, belonging to the structural classifications of phenols, polyphenols, ketones, aldehydes, and acids. For research use only, it is produced by commensal microbiota and has been shown to promote chemotaxis and virulence gene expression in Enterohemorrhagic Escherichia coli.

  • Endogenous metabolite of norepinephrine
  • High purity, 98.0%
  • Molecular weight of 184.15
  • Chemical formula C8H8O5
  • Appears as a white to off-white solid
  • Soluble in water and DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.