Learn More
Medchemexpress LLC 3,4-dihydroxymandelic acid | 775-01-9 | MFCD00004231 | >98.0% | 184.15 | C8H8O5 | 5 MG
SDP
Catalog No. 5000263980
Encompass_Preferred
3,4-Dihydroxymandelic acid is an endogenous metabolite of norepinephrine commonly used as an analytical standard in biochemical and metabolic studies. Supplied as a high-purity solid, it is appropriate for method development, calibration, and quantification of catecholamine metabolites in biological samples.
  • Used as an analytical standard for quantification of norepinephrine metabolites.
  • High purity (≥98.0%) supports accurate analytic results.
  • Chemical formula C8H8O5; molecular weight 184.15 g/mol.
  • White to off-white solid suitable for preparation of standards and controls.
  • Stable under frozen storage to preserve integrity for analytical use.
Catalog No. 50-002-63980 Supplier Medchemexpress LLC Supplier No. HY1134745MG
3,4-Dihydroxymandelic acid is an endogenous metabolite of norepinephrine commonly used as an analytical standard in biochemical and metabolic studies. Supplied as a high-purity solid, it is appropriate for method development, calibration, and quantification of catecholamine metabolites in biological samples.
  • Used as an analytical standard for quantification of norepinephrine metabolites.
  • High purity (≥98.0%) supports accurate analytic results.
  • Chemical formula C8H8O5; molecular weight 184.15 g/mol.
  • White to off-white solid suitable for preparation of standards and controls.
  • Stable under frozen storage to preserve integrity for analytical use.