Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3',4'-dimethoxy-7-hydroxyisoflavone

Catalog No. CDS007426 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS007426 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS007426 Supplier Sigma Aldrich Supplier No. CDS0074265MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

cis-11-Hexadecenal is the major pheromone component detected in the whole-animal male Helicoverpa zea antennal preparations.cis-11-Hexadecenal has been used in the preparation of fatty aldehyde semicarbazone derivatives during their analysis by liquid chromatography-electrospray ionization-tandem mass spectrometery.

Specifications

CAS 24160-14-3
Molecular Formula C17H14O5
Linear Formula C17H14O5
MDL Number MFCD00076013
Quantity 100 mg
Molecular Weight (g/mol) 298.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.